3-Fluorobenzal chloride ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: α,α-Dichloro-3-fluortoluene, 1-(Dichloromethyl)-3-fluorobenzene

L04121.06 L04121.14
ALFAL04121.06EA 194 SGD
ALFAL04121.06 ALFAL04121.14
3-Fluorobenzal chloride ≥98%
3-Fluorobenzal chloride

Purity: 98% CAS No.: 402-64-2 Molecular Formula: C7H5Cl2F Formula Weight: 179.02 Refractive Index: 1.5275 Boiling Point: 195-197° UN No.: 3265 MDL No.: MFCD00037112

Formula: C₇H₅Cl₂F
MW: 179,02 g/mol
Boiling Pt: 195…197 °C
Density: 1,36 g/cm³ (16 °C)
Storage Temperature: Ambient
MDL Number: MFCD00037112
CAS Number: 402-64-2
EINECS: 206-952-5
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR