2,3,4,5,6-Pentafluorotoluene ≥99%
Supplier: Thermo Scientific
|
Warning
|
B21419.09
B21419.18
ALFAB21419.09EA
186
SGD
ALFAB21419.09
ALFAB21419.18
2,3,4,5,6-Pentafluorotoluene ≥99%
2,3,4,5,6-Pentafluorotoluene
Purity: 99%
CAS No.: 771-56-2
Molecular Formula: C7H3F5
Formula Weight: 182.09
Flash Point: 34°(93°F)
Refractive Index: 1.4030
Melting Point: -30°
Boiling Point: 116-117°
Beilstein Reference: 1911885
UN No.: 1993
MDL No.: MFCD00000298
Formula:
C₇H₃F₅ MW: 182,09 g/mol Boiling Pt: 116…117 °C Melting Pt: –30 °C Density: 1,439 Flash Pt: 34 °C (93 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000298 CAS Number: 771-56-2 EINECS: 212-233-7 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...