α,α,α-Trifluoro-3,5-dinitrotoluene ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 3,5-Dinitrobenzotrifluoride

B24248.06 B24248.14
ALFAB24248.06EA 96.1 SGD
ALFAB24248.06 ALFAB24248.14
α,α,α-Trifluoro-3,5-dinitrotoluene ≥98%
α,α,α-Trifluoro-3,5-dinitrotoluene

Purity: 98% CAS No.: 401-99-0 Molecular Formula: C7H3F3N2O4 Formula Weight: 236.11 Flash Point: 110°(230°F) Melting Point: 50-52° Beilstein Reference: 2058079 MDL No.: MFCD00007233

Formula: C₇H₃F₃N₂O₄
MW: 236,11 g/mol
Boiling Pt: 127…129 °C (9 mmHg)
Melting Pt: 50…52 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007233
CAS Number: 401-99-0
EINECS: 206-935-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR