3-(Trifluoromethyl)anisole 98%

Supplier: Thermo Scientific
Warning

B25249.06 B25249.14
ALFAB25249.06EA 80.7 SGD
ALFAB25249.06 ALFAB25249.14
3-(Trifluoromethyl)anisole 98%
3-(Trifluoromethyl)anisole

Boiling Point: 159-160°
Beilstein Reference: 3048340

Formula: C₈H₇F₃O
MW: 176,14 g/mol
Boiling Pt: 159…160 °C
Melting Pt: –65 °C
Density: 1,215
Flash Pt: 48 °C (118 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000385
CAS Number: 454-90-0
EINECS: 207-229-7
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance (Colour) Clear colorless to pale yellow
Form Liquid
Assay (GC) ≥97.5%
Refractive Index 1.4420 - 1.4450 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR