sec-Butylbenzene ≥99%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-Phenylbutane
B22962.AC
B22962.AE
ALFAB22962.ACEA
0
SGD
ALFAB22962.AC
ALFAB22962.AE
sec-Butylbenzene ≥99%
sec-Butylbenzene
Purity: 99%
CAS No.: 135-98-8
Molecular Formula: C10H14
Formula Weight: 134.22
Flash Point: 52°(125°F)
Refractive Index: 1.4900
Odour: Aromatic
Melting Point: -75°
Boiling Point: 173-174°
Beilstein Reference: 1903902
Merck Reference: 14,1550
UN No.: 2709
MDL No.: MFCD00009329
Formula:
C₆H₅CH(CH₃)CH₂CH₃ MW: 134,22 g/mol Boiling Pt: 173…174 °C Melting Pt: –75 °C Density: 0,862 Flash Pt: 52 °C (125 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009329 CAS Number: 135-98-8 EINECS: 205-227-0 UN: 2709 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...