4'-Hydroxy-3'-nitroacetophenone ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
1-(4-Hydroxy-3-nitrophenyl)-1-ethanone
A18317.14
A18317.03
A18317.06
ALFAA18317.14EA
0
SGD
ALFAA18317.14
ALFAA18317.03
ALFAA18317.06
4'-Hydroxy-3'-nitroacetophenone ≥98%
4'-Hydroxy-3'-nitroacetophenone
Purity: 98%
CAS No.: 6322-56-1
Molecular Formula: C8H7NO4
Formula Weight: 181.15
Melting Point: 131-133°
Beilstein Reference: 1959078
Formula:
C₈H₇NO₄ MW: 181,15 g/mol Melting Pt: 131…133 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017002 CAS Number: 6322-56-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...