Fmoc-D-Pro-OH, (max. 5% H₂O) ≥98%

Supplier: Thermo Scientific

Synonyms: N-Fmoc-D-proline

H28500.14 H28500.06 H28500.03
ALFAH28500.14EA 1000 SGD
ALFAH28500.14 ALFAH28500.06 ALFAH28500.03
Fmoc-D-Pro-OH, (max. 5% H₂O) ≥98%
Fmoc-D-Pro-OH

Purity: 98+% CAS No.: 101555-62-8 Molecular Formula: C20H19NO4 Formula Weight: 337.38 Storage Temperature: 2°C to 8°C Melting Point: 110-116° MDL No.: MFCD00077067

Formula: C₂₀H₁₉NO₄
MW: 337,38 g/mol
Melting Pt: 110…116 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00077067
CAS Number: 101555-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR