2-Bromo-6-ethoxynaphthalene ≥98%

Supplier: Thermo Scientific
Warning

H50568.03 H50568.MD
ALFAH50568.03EA 505 SGD
ALFAH50568.03 ALFAH50568.MD
2-Bromo-6-ethoxynaphthalene ≥98%
2-Bromo-6-ethoxynaphthalene

Purity: 98% CAS No.: 66217-19-4 Molecular Formula: C12H11BrO Formula Weight: 251.12 MDL No.: MFCD01568865

Formula: C₁₂H₁₁BrO
Storage Temperature: Ambient
MDL Number: MFCD01568865
CAS Number: 66217-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR