(±)-4-Fluoro-α-methylbenzylamine ≥97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
DL-4-Fluoro-α-methylbenzylamine, 1-(4-Fluorophenyl)ethylamine
L04575.03
L04575.06
ALFAL04575.03EA
0
SGD
ALFAL04575.03
ALFAL04575.06
(±)-4-Fluoro-α-methylbenzylamine ≥97%
(±)-4-Fluoro-α-methylbenzylamine
Purity: 97%
CAS No.: 403-40-7
Molecular Formula: C8H10FN
Formula Weight: 139.17
Flash Point: 68°(154°F)
Refractive Index: 1.5020
Odour: Amine-like
pH: 10
Boiling Point: 76°/22mm
Beilstein Reference: 2802417
UN No.: 2735
MDL No.: MFCD00041323
Formula:
C₈H₁₀FN MW: 139,17 g/mol Boiling Pt: 76 °C (22 mmHg) Density: 1,030 Flash Pt: 68 °C (154 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041323 CAS Number: 403-40-7 EINECS: 206-958-8 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...