1-tert-Butyl-2-thiourea ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
N-(tert-Butyl)thiourea
L11979.06
L11979.03
ALFAL11979.06EA
0
SGD
ALFAL11979.06
ALFAL11979.03
1-tert-Butyl-2-thiourea ≥97%
1-tert-Butyl-2-thiourea
Purity: 97%
CAS No.: 7204-48-0
Molecular Formula: C5H12N2S
Formula Weight: 132.23
Odour: Odourless
Melting Point: 172-176°
Beilstein Reference: 1744506
MDL No.: MFCD00041192
Formula:
C₅H₁₂N₂S Melting Pt: 172…176 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041192 CAS Number: 7204-48-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...