8-Amino-1-naphthalenesulphonic acid 98%

Supplier: Thermo Scientific
Warning

Synonyms: 8-Aminonaphthalene-1-sulphonic acid

L17616.22 L17616.14
ALFAL17616.22EA 224 SGD
ALFAL17616.22 ALFAL17616.14
8-Amino-1-naphthalenesulphonic acid 98%
8-Amino-1-naphthalenesulphonic acid

Purity: 98% CAS No.: 82-75-7 Molecular Formula: C10H9NO3S Formula Weight: 223.25 Melting Point: 300° Beilstein Reference: 983230 Merck Reference: 14,6405 MDL No.: MFCD00035730

Formula: C₁₀H₉NO₃S
MW: 223,25 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00035730
CAS Number: 82-75-7
EINECS: 201-437-1
Merck Index: 14,6405

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR