(R)-(+)-4-Fluoro-α-methylbenzylamine 99%, ee 98%, ChiPros®
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
(R)-(+)-1-(4-Fluorophenyl)ethylamine, (R)-1-(4-Fluorophenyl)ethanamine
Purity: 99%
CAS No.: 374898-01-8
Molecular Formula: C8H10FN
Formula Weight: 139.17
Flash Point: 68°(154°F)
Refractive Index: 1.5020
Melting Point: -30°
Boiling Point: 76°/22mm
Beilstein Reference: 8614058
Optical Rotation: +33° (neat)
UN No.: 2735
MDL No.: MFCD03093090
Formula:
C₈H₁₀FN MW: 139,17 g/mol Boiling Pt: 76 °C (22 mmHg) Melting Pt: –30 °C Density: 1,030 Flash Pt: 68 °C (154 °F) Storage Temperature: Ambient |
MDL Number:
MFCD03093090 CAS Number: 374898-01-8 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...