(S)-(-)-4-Bromo-α-methylbenzylamine 99%, ee 98%, ChiPros®
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
(S)-1-(4-Bromophenyl)ethanamine, (S)-(-)-1-(4-Bromophenyl)ethylamine
L20080.03
L20080.06
L20080.14
ALFAL20080.03EA
176
SGD
ALFAL20080.03
ALFAL20080.06
ALFAL20080.14
(S)-(-)-4-Bromo-α-methylbenzylamine 99%, ee 98%, ChiPros®
(S)-(-)-4-Bromo-α-methylbenzylamine
Purity: 99%
CAS No.: 27298-97-1
Molecular Formula: C8H10BrN
Formula Weight: 200.08
Flash Point:
Formula:
C₈H₁₀BrN MW: 200,08 g/mol Boiling Pt: 140…145 °C (30 mmHg) Melting Pt: –25 °C Density: 1,371 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00066026 CAS Number: 27298-97-1 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...