3-(Methylthio)phenylboronic acid 97%

Supplier: Thermo Scientific
Warning

Synonyms: 3-Thioanisoleboronic acid, 3-(Methylthio)benzeneboronic acid

L20250.03 L20250.06 L20250.MD
ALFAL20250.03EA 0 SGD
ALFAL20250.03 ALFAL20250.06 ALFAL20250.MD
3-(Methylthio)phenylboronic acid 97%
3-(Methylthio)phenylboronic acid

Purity: 97% CAS No.: 128312-11-8 Molecular Formula: C7H9BO2S Formula Weight: 168.02 Melting Point: 158-161° Beilstein Reference: 7020728

Formula: C₇H₉BO₂S
MW: 168,02 g/mol
Melting Pt: 158…161 °C
Storage Temperature: Ambient
MDL Number: MFCD01319105
CAS Number: 128312-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR