2,5-Dimethylanisole ≥98%
Supplier: Thermo Scientific
Warning
|
Synonyms:
2-Methoxy-1,4-dimethylbenzene
B20456.14
B20456.22
ALFAB20456.14EA
0
SGD
ALFAB20456.14
ALFAB20456.22
2,5-Dimethylanisole ≥98%
2,5-Dimethylanisole
Purity: 98+%
CAS No.: 1706-11-2
Molecular Formula: C9H12O
Formula Weight: 136.19
Flash Point: 66°(150°F)
Refractive Index: 1.5140
Boiling Point: 189-190°
Beilstein Reference: 774604
MDL No.: MFCD00008378
Formula:
C₉H₁₂O MW: 136,19 g/mol Boiling Pt: 189…190 °C Density: 0,965 Flash Pt: 66 °C (150 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008378 CAS Number: 1706-11-2 EINECS: 216-943-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...