1-tert-Butyl-3,4,5-trimethylbenzene ≥97%

Supplier: Thermo Scientific

Synonyms: 5-tert-Butyl-1,2,3-trimethylbenzene

L05943.09 L05943.18
ALFAL05943.09EA 0 SGD
ALFAL05943.09 ALFAL05943.18
1-tert-Butyl-3,4,5-trimethylbenzene ≥97%
1-tert-Butyl-3,4,5-trimethylbenzene

Purity: 97% CAS No.: 98-23-7 Molecular Formula: C13H20 Formula Weight: 176.29 Melting Point: 31-33° Boiling Point: 114-115°/17mm MDL No.: MFCD00043642

Formula: C₁₃H₂₀
MW: 176,3 g/mol
Boiling Pt: 114…115 °C (17 mmHg)
Melting Pt: 31…33 °C
Storage Temperature: Ambient
MDL Number: MFCD00043642
CAS Number: 98-23-7
EINECS: 202-648-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR