8-Quinolinecarboxylic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Quinoline-8-carboxylic acid
L19644.03
L19644.MD
ALFAL19644.03EA
0
SGD
ALFAL19644.03
ALFAL19644.MD
8-Quinolinecarboxylic acid ≥98%
8-Quinolinecarboxylic acid
Purity: 98+%
CAS No.: 86-59-9
Molecular Formula: C10H7NO2
Formula Weight: 173.17
Melting Point: 185-189°
Beilstein Reference: 19176
Merck Reference: 14,8070
MDL No.: MFCD00047619
Formula:
C₁₀H₇NO₂ MW: 173,17 g/mol Melting Pt: 185…189 °C Storage Temperature: Ambient |
MDL Number:
MFCD00047619 CAS Number: 86-59-9 Merck Index: 14,8070 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...