Dicarbonylbis(triphenylphosphine)nickel ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: Bis(triphenylphosphine)nickel dicarbonyl, Bis(triphenylphosphine)dicarbonylnickel

39242.09 39242.18 39242.27
ALFA39242.09EA 176 SGD
ALFA39242.09 ALFA39242.18 ALFA39242.27
Dicarbonylbis(triphenylphosphine)nickel ≥98%
Dicarbonylbis(triphenylphosphine)nickel

Purity: 98% CAS No.: 13007-90-4 Molecular Formula: [(C6H5)3P]2Ni(CO)2 Formula Weight: 639.29 Physical Form: Crystalline Melting Point: 210-215° dec. UN No.: 3466 MDL No.: MFCD00009591

Formula: C₃₈H₃₀NiO₂P₂
MW: 639,29 g/mol
Melting Pt: 210…215 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00009591
CAS Number: 13007-90-4
EINECS: 235-851-9
UN: 3466
ADR: ,
Merck Index: 14,1304

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR