Bis(2-methoxyethyl)adipate ≥98%

Supplier: Thermo Scientific

Synonyms: Adipic acid bis(2-methoxyethyl) ester

A10497.14 A10497.22
ALFAA10497.14EA 0 SGD
ALFAA10497.14 ALFAA10497.22
Bis(2-methoxyethyl)adipate ≥98%
Bis(2-methoxyethyl) adipate

Purity: 98% CAS No.: 106-00-3 Molecular Formula: C12H22O6 Formula Weight: 262.30 Refractive Index: 1.4425 Boiling Point: 190°/11mm MDL No.: MFCD00041905

Formula: C₁₂H₂₂O₆
MW: 262,3 g/mol
Boiling Pt: 190 °C (11 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00041905
CAS Number: 106-00-3
EINECS: 203-352-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR