2-Chloro-4-nitrobenzonitrile ≥98%
Supplier: Thermo Scientific
|
Warning
|
A11977.03
A11977.06
ALFAA11977.03EA
0
SGD
ALFAA11977.03
ALFAA11977.06
2-Chloro-4-nitrobenzonitrile ≥98%
2-Chloro-4-nitrobenzonitrile
Purity: 98%
CAS No.: 28163-00-0
Molecular Formula: C7H3ClN2O2
Formula Weight: 182.57
Melting Point: 78-82°
MDL No.: MFCD00052203
Formula:
C₇H₃ClN₂O₂ Melting Pt: 78…82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052203 CAS Number: 28163-00-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...