Dihexyl phthalate ≥97%

Supplier: Thermo Scientific
Danger

A12593.09 A12593.30
ALFAA12593.09EA 0 SGD
ALFAA12593.09 ALFAA12593.30
Dihexyl phthalate ≥97%
Dihexyl phthalate

Purity: 97% CAS No.: 84-75-3 Molecular Formula: C20H30O4 Formula Weight: 334.46 Refractive Index: 1.4870 Boiling Point: 185-187°/0.5mm UN No.: 3082 MDL No.: MFCD00043720

Formula: C₂₀H₃₀O₄
Boiling Pt: 185…187 °C (0,5 mmHg)
Density: 1,01
Storage Temperature: Ambient
MDL Number: MFCD00043720
CAS Number: 84-75-3
EINECS: 201-559-5
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR