2-Amino-3,5-dibromo-6-methylpyridine 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
3,5-Dibromo-6-methyl-2-pyridinamine
B20498.03
B20498.06
ALFAB20498.03EA
0
SGD
ALFAB20498.03
ALFAB20498.06
2-Amino-3,5-dibromo-6-methylpyridine 97%
2-Amino-3,5-dibromo-6-methylpyridine
Purity: 97%
CAS No.: 91872-10-5
Molecular Formula: C6H6Br2N2
Formula Weight: 265.95
Melting Point: 144-145°
Beilstein Reference: 121839
Formula:
C₆H₆Br₂N₂ MW: 265,94 g/mol Melting Pt: 144…145 °C Storage Temperature: Ambient |
MDL Number:
MFCD00068229 CAS Number: 91872-10-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...