Chloro(di-tert-butyl)phosphine 96%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Di-tert-butylchlorophosphine, Di-t-butylchlorophosphine
B20998.06
B20998.14
ALFAB20998.06EA
0
SGD
ALFAB20998.06
ALFAB20998.14
Chloro(di-tert-butyl)phosphine 96%
Chloro(di-tert-butyl)phosphine
Purity: 96%
CAS No.: 13716-10-4
Molecular Formula: C8H18ClP
Formula Weight: 180.66
Flash Point: 62°(144°F)
Refractive Index: 1.4840
Odour: Acrid
Boiling Point: 47-49°/3mm
UN No.: 3265
MDL No.: MFCD00008815
Formula:
C₈H₁₈ClP MW: 180,66 g/mol Boiling Pt: 47…49 °C (3 mmHg) Density: 0,951 Flash Pt: 62 °C (144 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008815 CAS Number: 13716-10-4 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...