2,3-Dimethylpiperazine cis- and trans-mixture 95%

Supplier: Thermo Scientific
Warning

H26252.MD H26252.03
ALFAH26252.MDEA 0 SGD
ALFAH26252.MD ALFAH26252.03
2,3-Dimethylpiperazine cis- and trans-mixture 95%
2,3-Dimethylpiperazine

Purity: 95% CAS No.: 84468-52-0 Molecular Formula: C6H14N2 Formula Weight: 114.19 Melting Point: 81-90° MDL No.: MFCD07373397

Formula: C₆H₁₄N₂
MW: 114,19 g/mol
Melting Pt: 81…90 °C
Storage Temperature: Ambient
MDL Number: MFCD07373397
CAS Number: 84468-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR