3-Formyl-6-methoxypyridine ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
6-Methoxypyridine-3-carbaldehyde, 6-Methoxynicotinaldehyde, 6-Methoxypyridine-3-carboxaldehyde, 2-Methoxypyridine-5-carboxaldehyde, 6-Methoxy-3-pyridinecarboxaldehyde
H26515.03
H26515.06
ALFAH26515.03EA
208
SGD
ALFAH26515.03
ALFAH26515.06
3-Formyl-6-methoxypyridine ≥98%
3-Formyl-6-methoxypyridine
Purity: 98%
CAS No.: 65873-72-5
Molecular Formula: C7H7NO2
Formula Weight: 137.14
Flash Point: 107°(225°F)
Melting Point: 49-52°
MDL No.: MFCD02683446
Formula:
C₇H₇NO₂ MW: 137,14 g/mol Melting Pt: 49…52 °C Flash Pt: 107 °C (225 °F) Storage Temperature: Ambient |
MDL Number:
MFCD02683446 CAS Number: 65873-72-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...