2-(Boc-amino)pyridine 95%

Supplier: Thermo Scientific
Warning

Synonyms: tert-Butyl N-(2-pyridyl)carbamate, tert-Butyl pyridin-2-ylcarbamate

H30741.03 H30741.06
ALFAH30741.03EA 0 SGD
ALFAH30741.03 ALFAH30741.06
2-(Boc-amino)pyridine 95%
2-(Boc-amino)pyridine

Purity: 95% CAS No.: 38427-94-0 Molecular Formula: C10H14N2O2 Formula Weight: 194.23 Melting Point: 92-94° MDL No.: MFCD03411622

Formula: C₁₀H₁₄N₂O₂
MW: 194,23 g/mol
Melting Pt: 92…94 °C
Storage Temperature: Ambient
MDL Number: MFCD03411622
CAS Number: 38427-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR