2,6-Difluoro-4-methoxypyridine ≥97%
Supplier: Thermo Scientific
|
Warning
|
H34330.03
H34330.MD
ALFAH34330.03EA
0
SGD
ALFAH34330.03
ALFAH34330.MD
2,6-Difluoro-4-methoxypyridine ≥97%
2,6-Difluoro-4-methoxypyridine
Purity: 97%
CAS No.: 1184172-35-7
Molecular Formula: C6H5F2NO
Formula Weight: 145.11
Refractive Index: 1.6100
Formula:
C₆H₅F₂NO Storage Temperature: Ambient |
MDL Number:
MFCD13185564 CAS Number: 1184172-35-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...