3-Amino-4-formylpyridine 95%

Supplier: Thermo Scientific
Warning

Synonyms: 3-Amino-pyridine-4-carbaldehyde, 3-Aminoisonicotinaldehyde, 3-Aminopyridine-4-carboxaldehyde

H50024.03 H50024.MD
ALFAH50024.03EA 314 SGD
ALFAH50024.03 ALFAH50024.MD
3-Amino-4-formylpyridine 95%
3-Amino-4-formylpyridine

Purity: 95% CAS No.: 55279-29-3 Molecular Formula: C6H6N2O Formula Weight: 122.13 Melting Point: ca 92° MDL No.: MFCD06410684

Formula: C₆H₆N₂O
MW: 122,13 g/mol
Melting Pt: ∼ 92 °C
Storage Temperature: Ambient
MDL Number: MFCD06410684
CAS Number: 55279-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR