4-(Boc-amino)-2-chloropyridine 95%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Chloro-4-(tert-butoxycarbonyl)pyridine, tert-Butyl (2-chloropyridin-4-yl)carbamate, 4-Amino-2-chloropyridine, 4-BOC protected, tert-Butyl 2-chloropyridine-4-carbamate

H50067.03 H50067.06
ALFAH50067.03EA 0 SGD
ALFAH50067.03 ALFAH50067.06
4-(Boc-amino)-2-chloropyridine 95%
4-(Boc-amino)-2-chloropyridine

Purity: 95% CAS No.: 234108-73-7 Molecular Formula: C10H13ClN2O2 Formula Weight: 228.68 MDL No.: MFCD02179232

Formula: C₁₀H₁₃ClN₂O₂
MW: 228,68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02179232
CAS Number: 234108-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR