2-Acetyl-5-nitrobenzofuran ≥98%
Supplier: Thermo Scientific
L06308.03
L06308.06
ALFAL06308.03EA
0
SGD
ALFAL06308.03
ALFAL06308.06
2-Acetyl-5-nitrobenzofuran ≥98%
2-Acetyl-5-nitrobenzofuran
Purity: 98%
CAS No.: 23136-39-2
Molecular Formula: C10H7NO4
Formula Weight: 205.18
Melting Point: 173-177°
Beilstein Reference: 1684589
Formula:
C₁₀H₇NO₄ MW: 205,17 g/mol Melting Pt: 173…177 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051634 CAS Number: 23136-39-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...