2-Bromo-6-formylpyridine ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 6-Bromopyridine-2-carboxaldehyde, 6-Bromopyridine-2-carbaldehyde, 6-Bromo-2-pyridinecarboxaldehyde

L19518.03 L19518.06 L19518.14
ALFAL19518.03EA 123 SGD
ALFAL19518.03 ALFAL19518.06 ALFAL19518.14
2-Bromo-6-formylpyridine ≥97%
2-Bromo-6-formylpyridine

Purity: 97% CAS No.: 34160-40-2 Molecular Formula: C6H4BrNO Formula Weight: 186.01 Melting Point: 79-83° Beilstein Reference: 1524300

Formula: C₆H₄BrNO
MW: 186,01 g/mol
Melting Pt: 79…83 °C
Storage Temperature: Ambient
MDL Number: MFCD02683546
CAS Number: 34160-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR