2-Bromo-4-formylpyridine ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-Bromo-4-pyridinecarboxaldehyde, 2-Bromopyridine-4-carboxaldehyde
L19852.03
L19852.06
ALFAL19852.03EA
0
SGD
ALFAL19852.03
ALFAL19852.06
2-Bromo-4-formylpyridine ≥97%
2-Bromo-4-formylpyridine
Purity: 97%
CAS No.: 118289-17-1
Molecular Formula: C6H4BrNO
Formula Weight: 186.01
Storage Temperature: 2°C to 8°C
Melting Point: 52-56°
Beilstein Reference: 4243166
MDL No.: MFCD04039311
Formula:
C₆H₄BrNO MW: 186,01 g/mol Melting Pt: 52…56 °C |
MDL Number:
MFCD04039311 CAS Number: 118289-17-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...