2-Acetamido-5-aminopyridine 98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
2-(Acetylamino)-5-aminopyridine
L20004.03
L20004.06
ALFAL20004.03EA
116
SGD
ALFAL20004.03
ALFAL20004.06
2-Acetamido-5-aminopyridine 98%
2-Acetamido-5-aminopyridine
Purity: 98%
CAS No.: 29958-14-3
Molecular Formula: C7H9N3O
Formula Weight: 151.17
Melting Point: 144-154°
Beilstein Reference: 128346
Formula:
C₇H₉N₃O MW: 151,17 g/mol Melting Pt: 144…154 °C Storage Temperature: Ambient |
MDL Number:
MFCD00160311 CAS Number: 29958-14-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...