Rubidium nitrate ≥99.8% (metals basis)

Supplier: Thermo Scientific
Danger

12889.09 12889.18 12889.30
ALFA12889.09EA 0 SGD
ALFA12889.09 ALFA12889.18 ALFA12889.30
Rubidium nitrate ≥99.8% (metals basis)
Rubidium nitrate

Purity: 99.8% CAS No.: 13126-12-0 Molecular Formula: RbNO3 Formula Weight: 147.47 Physical Form: Crystalline Density: 3.11 Refractive Index: 1.52 Sensitivity: Hygroscopic Appearance: White Melting Point: 310° UN No.: 1477 MDL No.: MFCD00011193

Formula: NO₃Rb
MW: 147,47 g/mol
Boiling Pt: 578 °C (1013 hPa)
Melting Pt: 310 °C
Density: 3,11
Storage Temperature: Ambient
MDL Number: MFCD00011193
CAS Number: 13126-12-0
EINECS: 236-060-1
UN: 1477
ADR: 5.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR