Sodium niobate ≥99.997% (metals basis), Puratronic®

Supplier: Thermo Scientific
Warning

10872.06 10872.14
ALFA10872.06EA 0 SGD
ALFA10872.06 ALFA10872.14
Sodium niobate ≥99.997% (metals basis), Puratronic®
Sodium niobate

Purity: 99.997% CAS No.: 12034-09-2 Molecular Formula: NaNbO3 Formula Weight: 163.89 Physical Form: Powder Appearance: White to off-white Odour: Odourless Melting Point: 1422° MDL No.: MFCD00014230

Formula: NaNbO₃
Melting Pt: 1422 °C
Density: 4,42 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00014230
CAS Number: 12034-09-2
EINECS: 234-798-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR