Bis(cyclopentadienyl)manganese, sublimated
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Manganocene
ampouled under argon
14535.03
14535.06
ALFA14535.03EA
0
SGD
ALFA14535.03
ALFA14535.06
Bis(cyclopentadienyl)manganese, sublimated
Bis(cyclopentadienyl)manganese
CAS No.: 73138-26-8
Molecular Formula: C10H10Mn
Formula Weight: 185.13
Flash Point: 52°(126°F)
Physical Form: Powder, Ampouled under argon
Sensitivity: Air and Moisture Sensitive
Boiling Point: 245°
UN No.: 3181
MDL No.: MFCD00045468
Formula:
C₁₀H₁₀Mn MW: 185,13 g/mol Boiling Pt: 245 °C Melting Pt: 175 °C Flash Pt: 52 °C (126 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00045468 CAS Number: 73138-26-8 EINECS: 215-038-5 UN: 3181 ADR: 4.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...