Ethyl picolinate ≥99%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Ethyl 2-pyridinecarboxylate, Ethyl 2-picolinate
A13973.14
A13973.18
A13973.22
A13973.30
ALFAA13973.14EA
0
SGD
ALFAA13973.14
ALFAA13973.18
ALFAA13973.22
ALFAA13973.30
Ethyl picolinate ≥99%
Ethyl picolinate
Purity: 99%
CAS No.: 2524-52-9
Molecular Formula: C8H9NO2
Formula Weight: 151.17
Flash Point: 107°(224°F)
Refractive Index: 1.5105
Odour: Characteristic
Melting Point: 1-3°
Boiling Point: 239-241°
Beilstein Reference: 122485
MDL No.: MFCD00006292
Formula:
C₈H₉NO₂ MW: 151,16 g/mol Boiling Pt: 239…241 °C Melting Pt: 1…3 °C Density: 1,120 Flash Pt: 107 °C (224 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006292 CAS Number: 2524-52-9 EINECS: 219-758-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...