Lead tetraacetate 96% (dry weight) stabilised

Supplier: Thermo Scientific
Danger

Synonyms: Lead(IV) tetraacetate, Triacetyloxyplumbyl acetate, Acetic acid lead (IV) salt, Lead (IV) acetate, LTA

Stabilization: Moistured with glacial acetic acid 5-10%

A15551.0B A15551.18 A15551.22 A15551.30
ALFAA15551.0BEA 0 SGD
ALFAA15551.0B ALFAA15551.18 ALFAA15551.22 ALFAA15551.30
Lead tetraacetate 96% (dry weight) stabilised
Lead tetraacetate

Purity: 96% CAS No.: 546-67-8 Molecular Formula: C8H12O8Pb Formula Weight: 443.37 Melting Point: 175° Beilstein Reference: 3595640 Merck Reference: 14,5423 UN No.: 1616 MDL No.: MFCD00008693

Formula: (H₃CCOO)₄Pb
MW: 443,38 g/mol
Melting Pt: 175 °C
Density: 2,28
Storage Temperature: Ambient
MDL Number: MFCD00008693
CAS Number: 546-67-8
EINECS: 208-908-0
UN: 1616
ADR: 6.1,III
Merck Index: 14,5423

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR