Methyl-4-aminonicotinate 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Methyl 4-aminonicotinate

B25324.03 B25324.06
ALFAB25324.03EA 0 SGD
ALFAB25324.03 ALFAB25324.06
Methyl-4-aminonicotinate 98%
Methyl 4-aminonicotinate

Purity: 98% CAS No.: 16135-36-7 Molecular Formula: C7H8N2O2 Formula Weight: 152.15 Melting Point: 172-174° MDL No.: MFCD00955996

Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Melting Pt: 172…174 °C
Storage Temperature: Ambient
MDL Number: MFCD00955996
CAS Number: 16135-36-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR