(±)-4-Fluorostyrene oxide ≥98%
Supplier: Thermo Scientific
|
Warning
|
H26600.06
H26600.14
ALFAH26600.06EA
0
SGD
ALFAH26600.06
ALFAH26600.14
(±)-4-Fluorostyrene oxide ≥98%
(±)-4-Fluorostyrene oxide
Purity: 98%
CAS No.: 18511-62-1
Molecular Formula: C8H7FO
Formula Weight: 138.14
Flash Point: 77°(171°F)
Refractive Index: 1.508
Odour: Pungent
Boiling Point: 92°/14mm
UN No.: 2810
MDL No.: MFCD00043994
Formula:
C₈H₇FO MW: 138,14 g/mol Boiling Pt: 92 °C (14 mmHg) Density: 1,167 Flash Pt: 77 °C (171 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00043994 CAS Number: 18511-62-1 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...