2,6-Dichloro-4-nitropyridine ≥97%
Supplier: Thermo Scientific
|
Danger
|
H27131.06
H27131.03
ALFAH27131.06EA
0
SGD
ALFAH27131.06
ALFAH27131.03
2,6-Dichloro-4-nitropyridine ≥97%
2,6-Dichloro-4-nitropyridine
Purity: 97%
CAS No.: 25194-01-8
Molecular Formula: C5H2Cl2N2O2
Formula Weight: 192.99
Melting Point: 94-98°
MDL No.: MFCD05670545
Formula:
C₅H₂Cl₂N₂O₂ MW: 192,99 g/mol Melting Pt: 94…98 °C Storage Temperature: Ambient |
MDL Number:
MFCD05670545 CAS Number: 25194-01-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...