6-Bromopicolinic acid ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 6-Bromo-2-pyridinecarboxylic acid, 6-Bromopyridine-2-carboxylic acid

H30553.03 H30553.06
ALFAH30553.03EA 109 SGD
ALFAH30553.03 ALFAH30553.06
6-Bromopicolinic acid ≥97%
6-Bromopicolinic acid

Purity: 97% CAS No.: 21190-87-4 Molecular Formula: C6H4BrNO2 Formula Weight: 202.01 Melting Point: ca 188° dec. MDL No.: MFCD00093197

Formula: C₆H₄BrNO₂
MW: 202,01 g/mol
Melting Pt: ∼ 188 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00093197
CAS Number: 21190-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR