4-Bromopyridine-2-carbonitrile ≥97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
4-Bromo-2-cyanopyridine
H34424.03
H34424.MD
ALFAH34424.03EA
271
SGD
ALFAH34424.03
ALFAH34424.MD
4-Bromopyridine-2-carbonitrile ≥97%
4-Bromopyridine-2-carbonitrile
Purity: 97%
CAS No.: 62150-45-2
Molecular Formula: C6H3BrN2
Formula Weight: 183.01
Melting Point: 87-91°
UN No.: 3439
MDL No.: MFCD04065805
Formula:
C₆H₃BrN₂ Melting Pt: 87…91 °C Storage Temperature: Ambient |
MDL Number:
MFCD04065805 CAS Number: 62150-45-2 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...