2-Chloro-6-methoxyisonicotinic acid 96%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Chloro-6-methoxypyridine-4-carboxylic acid

H50168.03 H50168.06
ALFAH50168.03EA 0 SGD
ALFAH50168.03 ALFAH50168.06
2-Chloro-6-methoxyisonicotinic acid 96%
2-Chloro-6-methoxyisonicotinic acid

Purity: 96% CAS No.: 15855-06-8 Molecular Formula: C7H6ClNO3 Formula Weight: 187.58 Boiling Point: 209-213° MDL No.: MFCD00173928

Formula: C₇H₆ClNO₃
MW: 187,58 g/mol
Boiling Pt: 209…213 °C
Storage Temperature: Ambient
MDL Number: MFCD00173928
CAS Number: 15855-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR