Methyl-6-methylnicotinate ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: Methyl 6-methylnicotinate

L12276.06 L12276.14 L12276.03
ALFAL12276.06EA 0 SGD
ALFAL12276.06 ALFAL12276.14 ALFAL12276.03
Methyl-6-methylnicotinate ≥97%
Methyl 6-methylnicotinate

Purity: 97+% CAS No.: 5470-70-2 Molecular Formula: C8H9NO2 Formula Weight: 151.17 Flash Point: 103°(217°F) Melting Point: 31-36° Boiling Point: 152-154°/40mm Beilstein Reference: 122940 MDL No.: MFCD00006340

Formula: C₈H₉NO₂
MW: 151,16 g/mol
Boiling Pt: 152…154 °C (40 mmHg)
Melting Pt: 31…36 °C
Flash Pt: 103 °C (217 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006340
CAS Number: 5470-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR