2,6-Dichloro-5-fluoronicotinic acid 96%

Supplier: Thermo Scientific
Warning

Synonyms: 2,6-Dichloro-5-fluoropyridine-3-carboxylic acid

L19454.14 L19454.06
ALFAL19454.14EA 0 SGD
ALFAL19454.14 ALFAL19454.06
2,6-Dichloro-5-fluoronicotinic acid 96%
2,6-Dichloro-5-fluoronicotinic acid

Purity: 96% CAS No.: 82671-06-5 Molecular Formula: C6H2Cl2FNO2 Formula Weight: 209.99 Melting Point: 152-155° Beilstein Reference: 4311035

Formula: C₆H₂Cl₂FNO₂
MW: 209,99 g/mol
Melting Pt: 152…155 °C
Storage Temperature: Ambient
MDL Number: MFCD00799517
CAS Number: 82671-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR