5-Bromo-2-nitropyridine 98

Supplier: Thermo Fisher Scientific
Warning

L19748.03 L19748.06
ALFAL19748.03EA 0 SGD
ALFAL19748.03 ALFAL19748.06
5-Bromo-2-nitropyridine 98
5-Bromo-2-nitropyridine

Purity: 98+% CAS No.: 39856-50-3 Molecular Formula: C5H3BrN2O2 Formula Weight: 203.00 Melting Point: 147-151° Beilstein Reference: 120878

Formula: C₅H₃BrN₂O₂
MW: 203
Melting Pt: 147…151 °C
Storage Temperature: Ambient
MDL Number: MFCD00160411
CAS Number: 39856-50-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR