5-Chloro-2-hydroxy-3-nitropyridine ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 5-Chloro-3-nitropyridin-2-ol

L19860.03 L19860.06
ALFAL19860.03EA 0 SGD
ALFAL19860.03 ALFAL19860.06
5-Chloro-2-hydroxy-3-nitropyridine ≥97%
5-Chloro-2-hydroxy-3-nitropyridine

Purity: 97% CAS No.: 21427-61-2 Molecular Formula: C5H3ClN2O3 Formula Weight: 174.54 Melting Point: 234-236° dec. Beilstein Reference: 383852

Formula: C₅H₃ClN₂O₃
MW: 174,54 g/mol
Melting Pt: 234…236 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00114884
CAS Number: 21427-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR