2,3,5-Trifluoroisonicotinic acid ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2,3,5-Trifluoropyridine-4-carboxylic acid
L20050.03
L20050.06
L20050.MD
ALFAL20050.03EA
0
SGD
ALFAL20050.03
ALFAL20050.06
ALFAL20050.MD
2,3,5-Trifluoroisonicotinic acid ≥97%
2,3,5-Trifluoroisonicotinic acid
Purity: 97%
CAS No.: 675602-91-2
Molecular Formula: C6H2F3NO2
Formula Weight: 177.08
Melting Point: 108-112°
MDL No.: MFCD03840205
Formula:
C₆H₂F₃NO₂ MW: 177,08 g/mol Melting Pt: 108…112 °C Storage Temperature: Ambient |
MDL Number:
MFCD03840205 CAS Number: 675602-91-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...