5-Bromo-3-pyridineboronic acid tech. 85%, Technical Grade

Supplier: Thermo Scientific
Warning

Synonyms: 3-Bromopyridine-5-boronic acid, 3-Bromopyridyl-5-boronic acid

L20084.03 L20084.MD
ALFAL20084.03EA 312 SGD
ALFAL20084.03 ALFAL20084.MD
5-Bromo-3-pyridineboronic acid tech. 85%, Technical Grade
5-Bromo-3-pyridineboronic acid

Purity: 85% CAS No.: 452972-09-7 Molecular Formula: C5H5BBrNO2 Formula Weight: 201.82 Storage Temperature: -30°C to -10°C Melting Point: 300° Beilstein Reference: 9119873 MDL No.: MFCD02685634

Formula: C₅H₅BBrNO₂
MW: 201,81 g/mol
Melting Pt: >300 °C
Storage Temperature: Freezer
MDL Number: MFCD02685634
CAS Number: 452972-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR